Methyl green zinc chloride salt, dark powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:7114-03-6
:
Formula:C₂₇H₃₅BrClN₃·xZnCl₂
MDL Number:MFCD00151094
:653.24 g/mol
Specification Test Results
- Synonyms:CI 42590; 4-[[4-(Dimethylamino) phenyl][4-(dimethyliminio) cyclohexa-2,5-dien-1-ylidene] methyl]- N-ethyl-N,N- bromide chloride, compound with zinc chloride; Ethyl Green
- Form:Dark Powder
- Storage:Store at Room Temperature (15 - 30 °C)
- Color Index:42590
- Extinction Coefficient:E1% = 800 - 1100 (water) Lit.
- Typical Working Concentration:A typical working concentration for using methyl green as a counter stain is 0,1 - 0,5%
Specification Test Results
Frequently Bought Together