Order Entry
France
Orders LinkContactUsLinkComponent
PIG® Crysta-lite absorbent granules

PIG® Crysta-lite absorbent granules

Absorbent granules, 18 L, 1 bag
Catalog # NEWPP45-1
Supplier:   NEW PIG
PIG® Crysta-lite absorbent granules
Catalog # NEWPP45-1
Supplier:  NEW PIG
PIG® Crysta-lite absorbent granules
PIG® Crysta-lite absorbent granules

Absorbent granules, 18 L, 1 bag

Price
16,90 €
Log in for availability
1 * 1 Pce
Product Details & Documents

An economical, light weight, non-combustible loose absorbent for spillages of water, oils, organic solvents, and most acids. PIG® Crysta-lite Absorbent will absorb many times its own weight.

  • Can be used as an absorbent media in enclosed containers for the disposal of liquid toxic waste substances
  • Remains granular, even when saturated, to provide excellent retention; stands up to heavy traffic without releasing liquids
  • Material is quarried, crushed, processed and screened to create a particle size of 3.0mm to 6.0mm
  • The "expansion" of the product generates countless tiny air bubbles within the structure creating the ultra light weight of PIG® Crysta-lite Absorbent
  • High surface area and excellent, fast-absorption properties give Absorbent the ability to create a low-slip surface
  • Chemically inert product is ideal for the absorption and removal of chemical spills, oil and water spillages on floors, nuclear waste, hazardous chemicals and more
  • Non-toxic mineral product is safe to handle and use without any risk

Weight: 3,5 kg per bag

Volume bag: 30 L

Specifications
  • Description:
    Absorbent granules
  • Seq:
    01
  • Absorbency:
    18 L
  • Packed:
    1 bag
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.