Order Entry
France
ContactUsLinkComponent
Concrete inspection hammer
Concrete inspection hammer
Catalog # SIMELD0500
Supplier:  Industrial Physics
undefined
Concrete inspection hammer
Catalog # SIMELD0500
Supplier:  Industrial Physics
Specifications
  • Range:
    10 - 110 N/mm²
  • Description:
    Concrete inspection hammer
  • Pk:
    1
Specifications
About this item
Hammer used to test the quality of concrete, as well as other construction materials, for analysing resistance.
Certifications: EN 12504-2, ASTM, C805, BS 1881.