Order Entry
France
ContactUsLinkComponent
 
Anti-gD Mouse Monoclonal Antibody [clone: J357]
Catalog # ABNOMAB13575
Supplier:  Abnova
Anti-gD Mouse Monoclonal Antibody [clone: J357]
Catalog # ABNOMAB13575
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Herpes simplex virus type 1/2 gD
  • Antigen symbol:
    gD
  • Clonality:
    Monoclonal
  • Clone:
    J357
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Herpes simplex virus
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Mouse monoclonal antibody raised against herpes simplex virus type 1/2 (HSV-1/2) gD.

Type: Primary
Antigen: gD
Clonality: Monoclonal
Clone: J357
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Herpes simplex virus (HSV)