Chromatography tubing, stainless steel
Supplier: VICI JOUR RESEARCH
Certificates
About this item
These flexible pre-cut stainless steel tubings are made of ¹/₃₂" premium grade 316 stainless steel tubing with laser-welded ends into ¹/₁₆". Available in different lengths most commonly required in UHPLC and HPLC systems.
- For zero dead volume connections
- Steam-cleaned and ready to use
- Soft annealed OD for easy ferrule connections
- Color bands for quick identification
- Softener-free polyethylene caps are used to prevent any contamination
OD tolerance is ±0,002", while the ID tolerance is ±0,001".
Tubings can withstand a pressure rating of min 365 bar (5300 psi).
Frequently Bought Together