Order Entry
France
ContactUsLinkComponent
Aluminium potassium sulphate dodecahydrate 98-102% ACS
Aluminium potassium sulphate dodecahydrate 98-102% ACS
Catalog # N984-500G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Aluminium potassium sulphate dodecahydrate 98-102% ACS
Catalog # N984-500G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Ammonium:
    <0.005%
  • Assay:
    98 - 102%
  • Chloride:
    ≤5 ppm
  • Heavy Metals (as Pb):
    ≤0.001%
  • Insoluble Matter:
    ≤0.005%
  • Iron:
    ≤0.001%
  • Sodium:
    ≤0.02%

Specification Test Results

About this item

 :
 :474.39 g/mol
Merck Index:12,00373
Melting Pt:92.5 °C
Density:1.72 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-141-3
CAS Number:7784-24-9
Formula:AlK(SO₄)₂·12H₂O
MDL Number:MFCD00149143