

N-Boc-4-oxo-L-proline methyl ester 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:102195-80-2
:
Formula:C₁₁H₁₇NO₅
MDL Number:MFCD01861778
:243.259 g/mol
Melting Pt:42…45 °C
Flash Pt:>110 °C
:GHS05
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to tan Solid
- Infrared spectrum:Conforms
- Melting point:42 to 46 °C
- GC:≥96.0 %
- Specific optical rotation:+10° to +17° (20 °C, 589 nm) (c=1, CHCl3)
Specification Test Results
Frequently Bought Together