Mercury(II) acetate ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1600-27-7
:
Formula:(H₃CCOO)₂Hg
MDL Number:MFCD00012165
:318.68 g/mol
Melting Pt:179…182 °C
Density:3,27 g/cm³ (20 °C)
UN:1629
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Titration with NH4SCN:≥98.0 %
- Chloride (Cl):≤0.005 %
- Nitrate (NO3):≤0.005 %
- Sulfate (SO4):≤0.005 %
- Iron (Fe):≤0.001 %
- Other heavy metals (as Pb):≤0.002 %
- Mercurous Mercury (as Hg):≤0.4 %
- Insoluble matter:≤0.01 %
- Residue after reduction:≤0.02 %
Specification Test Results
Frequently Bought Together