Order Entry
France
ContactUsLinkComponent
Ethanol denatured 100%, Technical Grade (denatured with 5% methanol)
Ethanol denatured 100%, Technical Grade (denatured with 5% methanol)
Catalog # ACRO444160050
CAS Number:  
undefined
Ethanol denatured 100%, Technical Grade (denatured with 5% methanol)
Catalog # ACRO444160050
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    5 L
  • Seq:
    5190
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Refractive index:
    1.3590 to 1.3610 (20°C, 589 nm)
  • Density:
    (20°C) 0.788 to 0.790 kg/l
  • GC:
    ≤5 % (methanol)

Specification Test Results

About this item

Synonyms: Ethyl alcohol denatured
 :
 :46.07 g/mol
Boiling Pt:∼78 °C (1013 hPa)
Melting Pt:–117 °C
Density:0.8 g/cm³
Flash Pt:12 °C
UN:1170
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:64-17-5
Formula:H₃CCH₂OH
MDL Number:MFCD00003568