Order Entry
France
ContactUsLinkComponent
2,3-Diphenylmaleic anhydride 98%
Irritant
2,3-Diphenylmaleic anhydride 98%
Certificates
undefined
Irritant
2,3-Diphenylmaleic anhydride 98%

Specification Test Results

  • Appearance:
    Yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    156 to 162 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together