3-Amino-2-methylpyridine 97%
Catalog # ACRO437630010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to tan Powder
- Infrared spectrum:Conforms
- Melting point:114 to 119 °C
- GC:≥96.0 %
Specification Test Results
About this item
:
:108.14 g/mol
Melting Pt:116…118 °C
ADR:6.1,III
:GHS07
Storage Temperature:Ambient
CAS Number:3430-10-2
Formula:C₆H₈N₂
MDL Number:MFCD03788195