



(R)-(+)-2-Methyl-CBS-oxazaborolidine 1 M in toluene, AcroSeal®
Catalog # 436040500.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:50 mL
- Seq:2370
Specifications
Specification Test Results
- Appearance:Colorless to amber Liquid
- Strength:0.90 to 1.10 M
Specification Test Results
About this item
CAS Number:112022-83-0
:
Formula:C₁₈H₂₀BNO
MDL Number:MFCD00078440
:277.17 g/mol
Boiling Pt:111 °C (760 mmHg)
Density:0.95 g/cm³
Flash Pt:4 °C
UN:1294
ADR:3,II
:GHS08
:GHS05
:GHS02
:GHS07
Storage Temperature:Ambient