Order Entry
France
ContactUsLinkComponent
 
Health
Corrosive
Flammable
Irritant
(S)-(-)-2-Methyl-CBS-oxazaborolidine 1 M in toluene, AcroSeal®
Catalog # 436030500.
CAS Number:  
undefined
Health
Corrosive
Flammable
Irritant
(S)-(-)-2-Methyl-CBS-oxazaborolidine 1 M in toluene, AcroSeal®
Catalog # 436030500.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • Appearance:
    Colorless to amber Liquid
  • Strength:
    0.90 to 1.10 M
  • Titration Argentometric:
    ≥85.0 %

Specification Test Results

About this item

CAS Number:112022-81-8
 :
Formula:C₁₈H₂₀BNO
MDL Number:MFCD00078439
 :277.17 g/mol
Boiling Pt:111 °C (760 mmHg)
Density:0.929 g/cm³
Flash Pt:4 °C
UN:1294
ADR:3,II
 :GHS08
 :GHS05
 :GHS02
 :GHS07
Storage Temperature:Ambient