1,2-Bis((2R,5R)-2,5-diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate 97%
Catalog # ACRO435202500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:250 mg
- Seq:2590
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Yellow to orange to red to brown Powder
- Infrared spectrum:Conforms
- NMR (1H-NMR):Consistent with structure
- NMR (31P-NMR):Consistent with structure
- Elemental Analysis:54.7 to 59.3 % C
- Specific optical rotation:-78° to -86° (20°C, 589 nm) (c=0.8 in chloroform)
Specification Test Results
About this item
Synonyms: (+)-1,2-Bis((2R,5R)-2,5-di-i-propylphospholano)benzene)1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, (+)-1,2-Bis((2R,5R)-2,5-di-i-propylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, (+)-1,2-BIS((2R,5R)-2,5-DI-I-PROPYLPHOSPHOLANO)BENZENE)1,5-CYCLOOCTADIENE)RHODIUM (I) TETRAFLUOROBORATE, (R,R)-i-Pr-DUPHOS-Rh
:
:714.45 g/mol
Storage Temperature:Ambient
CAS Number:569650-64-2
Formula:C₃₄H₅₄BF₄P₂Rh
MDL Number:MFCD07369039