O-tert-Butyl-L-serine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18822-58-7
:
Formula:C₇H₁₅NO₃
MDL Number:MFCD00066088
Melting Pt:225.2…228 °C
:161.20 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Water:≤10.0 % (K.F.)
- HPLC:≥96.0 %
- Specific optical rotation:-15.5° to -17.5° (20°C, 589 nm)
Specification Test Results
Frequently Bought Together