

Kaempferol 97%
Catalog # ACRO434731000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Yellow Powder
- Infrared spectrum:Conforms
- Water:≤10 % (K.F.)
- HPLC:≥96.0 %
Specification Test Results
About this item
CAS Number:520-18-3
:
Formula:C₁₅H₁₀O₆
MDL Number:MFCD00016938
:286.24 g/mol
Melting Pt:282…286 °C
UN:2811
:GHS06
:GHS08
Storage Temperature:Refrigerator