Order Entry
France
ContactUsLinkComponent
Propanephosphonic acid cyclic anhydride 50% (w/w) in ethyl acetate, anhydrous, AcroSeal®
Irritant
Corrosive
Flammable
Propanephosphonic acid cyclic anhydride 50% (w/w) in ethyl acetate, anhydrous, AcroSeal®
Catalog # 429311000.
CAS Number:  
undefined
Irritant
Corrosive
Flammable
Propanephosphonic acid cyclic anhydride 50% (w/w) in ethyl acetate, anhydrous, AcroSeal®
Catalog # 429311000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to amber Solution
  • Assay:
    ≥50 % w/w Solids
  • Alkalinity:
    ≤0.03 % (as Ca(OH)2)
  • Phosphate (PO4):
    ≤0.003 %
  • Sulfate (SO4):
    ≤0.05 %
  • Iron (Fe):
    ≤0.005 %
  • Acidity (HCl):
    ≤0.025 %
  • Insoluble matter:
    ≤0.025 % (in water and ppt by NH4OH)

Specification Test Results

About this item

 :
 :318.19 g/mol
Boiling Pt:77.2 °C
Density:1.069 g/cm³
Flash Pt:–5 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
 :GHS07
 :GHS05
 :GHS02
CAS Number:68957-94-8
Formula:C₉H₂₁O₆P₃
MDL Number:MFCD00006583