Benzyl chloroformate 50% (w/w) 3 M in toluene, AcroSeal®
Catalog # 428901000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear light yellow to light pink Solution
- GC:45 to 53 % w/w
Specification Test Results
About this item
CAS Number:501-53-1
:
Formula:C₈H₇ClO₂
MDL Number:MFCD00000640
:170.6 g/mol
Density:1.01 g/cm³
Flash Pt:7 °C
Storage Temperature:Refrigerator
UN:2920
ADR:8,I
:GHS07
:GHS08
:GHS05
:GHS09
:GHS02