Glycine benzyl ester hydrochloride (H-Gly-OBzl.HCl) 97%
:2462-31-9
:
:C₉H₁₂ClNO₂
:MFCD00035442
:201.65 g/mol
:Ambient
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
- NMR:consistent with structure
- Iron (Fe):≤0.0001 %
- Heavy metals (as Pb):≤1 ppm
Frequently Bought Together