Order Entry
France
ContactUsLinkComponent
2-(Boc-amino)ethanethiol 97%
Irritant
2-(Boc-amino)ethanethiol 97%
Certificates
undefined
Irritant
2-(Boc-amino)ethanethiol 97%

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid or low melting solid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4740 to 1.4770 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together