



(S)-(-)-2-Methyl-CBS-oxazaborolidine 1 M in toluene
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112022-81-8
:
Formula:C₁₈H₂₀BNO
MDL Number:MFCD00078439
:277.17 g/mol
Boiling Pt:111 °C (760 mmHg)
Density:0.929 g/cm³
Flash Pt:4 °C
UN:1294
ADR:3,II
:GHS08
:GHS05
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to amber Solution
- Strength:0.95 to 1.10 M
Specification Test Results
Frequently Bought Together