Acetonitrile-[D₃] 99.8 atom % D, AcroSeal® for NMR spectroscopy
Catalog # 217421000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.03 % (Coulometric)
- D-Enrichment:≥99.75 %
Specification Test Results
About this item
:
:44.03 g/mol
Boiling Pt:79…81 °C (760 mmHg)
Melting Pt:–46 °C
Density:0.844 g/cm³ (20 °C)
Flash Pt:2 °C
UN:1648
ADR:3,II
:GHS02
:GHS07
Storage Temperature:Ambient
CAS Number:2206-26-0
Formula:D₃CCN
MDL Number:MFCD00001881