Order Entry
France
ContactUsLinkComponent
7-Methoxy-1-tetralone 97%
Irritant
7-Methoxy-1-tetralone 97%
Certificates
undefined
Irritant
7-Methoxy-1-tetralone 97%

Specification Test Results

  • Appearance:
    White to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    60 to 64 °C
  • GC:
    ≥96.0 %
  • Elemental Analysis:
    44.65 to 48.37 % (C)

Specification Test Results

Related Information

Frequently Bought Together