
Paraffin, liquid, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:8012-95-1
:8002-74-2
Boiling Pt:300 °C (760 mmHg)
Melting Pt:–24 °C
Density:0.88 g/cm³
Flash Pt:218 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Oily liquid
- Infrared spectrum:Conforms
- Refractive index:1.4700 to 1.4800 (20°C, 589 nm)
- Density:(20°C) 0.827 to 0.890 g/ml
- Viscosity:63 to 80 mm²/s (40°C)
Specification Test Results
Frequently Bought Together