VWR® Antibody signal enhancer
Catalog # M336-1L
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:1 L
- Description:Antibody Signal Enhancer, 1000 ml, for 50 blots
- Seq:03
Specifications
About this item
Antibody Signal Enhancer is a Western blotting antibody diluent that increases intensity of protein detection up to 5-fold compared to conventional blotting.
- Protein-free solution eliminates cross-reaction with antibodies
- Reduces antigen and reagent use without the need for additional incubation steps
- Formulated for use with HRP- and biotin-conjugated antibodies and works with both PVDF and nitrocellulose blots