HyClone™ LS1000 Lipid supplement
Catalog # HYCLSH30554.02
Supplier: HyClone products (Cytiva)
CAS Number:
Specifications
- Pk:100 mL
- Description:LS1000 Lipid Supplement, ADCF (liquid)
Specifications
About this item
HyClone™ LS1000 Lipid Supplement is a 1000X concentrated supplement that has been formulated using a proprietary complexing process for enhanced cholesterol delivery.
- Stimulates cell growth and monoclonal antibody production in cholesterol auxotrophic NS0 cell lines
- Developed through Metabolic Pathway Design
- Contains no animal-derived components (ADCF)
- May be used to supplement serum-free media and in the preparation of liquid media media from dry powdered media
- Delivers 2,5 mg of cholesterol per ml