Order Entry
France
ContactUsLinkComponent
Chloroform ≥99.8% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Chloroform ≥99.8% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Certificates
undefined
Chloroform ≥99.8% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Certificates

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Boiling point:
    60 to 62 °C
  • Density:
    (20/4 °C) 1.474 to 1.483
  • GC:
    ≥99.80 % (Excluding stabilizer)
  • Color scale:
    ≤10 APHA
  • Stabilizer:
    0.5 to 1.0 % Ethanol
  • Residue after evaporation:
    ≤0.001 %
  • Substance darkened by H2SO4:
    Passes test
  • Free chlorine:
    (Cl) Passes test
  • Lead (Pb):
    ≤0.05 ppm
  • Acid and chloride:
    Passes test
  • Acetone and aldehyde:
    Passes test

Specification Test Results

Related Information

Frequently Bought Together