Methyl p-tolyl sulphoxide 98%
:934-72-5
:
:C₈H₁₀OS
:MFCD00075175
:154.227 g/mol
:113…114 °C (2.2 mmHg)
:41…46 °C
:>110 °C
:Ambient
- Appearance:White to yellow Low melting powder or crystals
- Infrared spectrum:Conforms
- Melting point:41 to 46 °C
- GC:≥97.5 %
- NMR:consistent with structure
Frequently Bought Together