
Staurosporine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:62996-74-1
:
Formula:C₂₈H₂₆N₄O₃
MDL Number:MFCD18252446
:466.541 g/mol
Melting Pt:270 °C
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to light yellow to light beige Powder
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together