Phenyl β-D-glucopyranoside hydrate 98%
ACRO376890050
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:White to beige Crystalline powder or crystals
- Infrared spectrum:Conforms
- Water:≤9 % (K.F.)
- HPLC:≥97.5 %
- Specific optical rotation:-70° to -75° (20°C, 589 nm) (C=1, H2O) (On dry substance)
:
:256.255 g/mol (anhy)
:172…176 °C
:Refrigerator
:1266359-89-0
:636565-38-3
:C₁₂H₁₆O₆.xH₂O
:MFCD03410292