Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers) 95%
: Methylnadic Anhydride, Methylnadic anhydride, Nadic Methyl Anhydride, Methyl-3,6-endomethylenetetrahydrophthalic Anhydride
:25134-21-8
:
:C₁₀H₁₀O₃
:MFCD00167036
:178.19 g/mol
:275 °C (760 mmHg)
:1.23 g/cm³
:140 °C
:2810
:6.1,III
:GHS05
:GHS06
:GHS08
:Ambient
- Environmentally Preferable:
- Appearance:Clear light yellow Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.5060 to 1.5080 (20°C, 589 nm)
- GC:≥94 %
Frequently Bought Together