Poly(methyl methacrylate), M.W. ∼35000, beads
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9011-14-7
:
Formula:C₅H₈O₂([-CH₂C(CH₃)(CO₂CH₃)-]n)
MDL Number:MFCD00134349
Melting Pt:105 °C
Flash Pt:300 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Beads
- Infrared spectrum:Conforms
- Loss on drying:≤5 %
- Particle size:50 to 150 microns (approximately)
Specification Test Results
Frequently Bought Together