ɑ-Bromocinnamaldehyde 98%
Glass bottle, 250 g
Supplier: Thermo Fisher Scientific
ɑ-Bromocinnamaldehyde 98%
Catalog # ACRO171272500
Supplier: Thermo Fisher Scientific
CAS Number: 5443-49-2
Jump to:
Product Details & Documents
CAS No.: 5443-49-2
Synonyms: 2-Bromo-3-phenyl-2-propenal, 2-Bromo-3-phenylacrylaldehyde, Alpha-Bromocinnamaldehyde
- CAS Number:5443-49-2
- Formula:C₉H₇BrO
- MDL Number:MFCD00006965
- Molecular Weight:211.06 g/mol
- Melting Pt:68…72 °C
- Storage Temperature:Ambient
Documents
ACRO17127
SDS
Specifications
Specification Test Results
- Appearance:Light yellow to ochre Crystalline powder
- Infrared spectrum:Conforms
- Melting point:68 to 72 °C
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:250 g



