Order Entry
France
ContactUsLinkComponent
 
Diethyl-trans-1,2-cyclopropanedicarboxylate 97%
Catalog # ACRO169180050
CAS Number:  
undefined
Diethyl-trans-1,2-cyclopropanedicarboxylate 97%
Catalog # ACRO169180050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4390 to 1.4420 (20°C, 589 nm)
  • GC:
    ≥96.0 %
  • Elemental Analysis:
    25.34 to 27.45 % (N)

Specification Test Results

About this item

 :
 :186.207 g/mol
Boiling Pt:70…75 °C (1 mmHg)
Density:1.06 g/cm³
Flash Pt:98 °C
Storage Temperature:Ambient
CAS Number:3999-55-1
Formula:C₉H₁₄O₄
MDL Number:MFCD00198002