
Triethyl-4-phosphonocrotonate predominantly trans 92%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10236-14-3
:
Formula:C₁₀H₁₉O₅P
MDL Number:MFCD00009192
:250.231 g/mol
Boiling Pt:133…135 °C (.4 mmHg)
Density:1.12 g/cm³
Flash Pt:>112 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4525 to 1.4555 (20°C, 589 nm)
- GC:≥90 %
Specification Test Results
Frequently Bought Together