
(3R,4R)-1-Benzylpyrrolidine-3,4-diol 97%
ACRO348720050
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:Off-white to yellow to light brown Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:95 to 101 °C
- GC:≥96.0 %
- Specific optical rotation:-29° to -35° (20 °C, 589 nm) (c=1, MeOH)
:
:95…101 °C
:193.2423 g/mol
:GHS07
:Ambient
:163439-82-5
:C₁₁H₁₅NO₂
:MFCD01073894