Order Entry
France
ContactUsLinkComponent
4-Methylumbelliferyl-β-D-glucopyranoside 99%
4-Methylumbelliferyl-β-D-glucopyranoside 99%
Certificates
undefined
4-Methylumbelliferyl-β-D-glucopyranoside 99%

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤2 % (K.F.)
  • TLC:
    ≥98.5 %
  • Appearance of solution:
    (3% soln. in acetone/water (2:1)) Clear, colorless
  • Specific optical rotation:
    -97° to -101° (20°C, 589 nm) (c=0.3, water)

Specification Test Results

Related Information

Frequently Bought Together