Order Entry
France
Orders LinkContactUsLinkComponent
(S)-(-)-2-Methoxypropionic acid 98%

(S)-(-)-2-Methoxypropionic acid 98%

Corrosive
(S)-(-)-2-Methoxypropionic acid 98%
Certificates
(S)-(-)-2-Methoxypropionic acid 98%

2 Options Available Below

Product Details & Documents

Appearance: Clear colorless to light yellow Liquid

Synonyms:  (S)-2-Methoxypropanoic acid
  • CAS Number:23953-00-6
  • Formula:C₄H₈O₃
  • MDL Number:MFCD01632589
  • Molecular Weight:104.105 g/mol
  • Boiling Pt:99 °C (20 mmHg)
  • Flash Pt:87 °C
  • Storage Temperature:Ambient
  • UN:3265
  • ADR:8,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4132 to 1.4152 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Specific optical rotation:
    -70° to -80° (20°C, 589 nm) (neat)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 177,00 €
    177,00 €
    1 * 500 mg
       177,00 €

  • Glass bottle
    2,5 g
    Log in for availability
    Log in for availability
    1 * 2,5 g
    1 * 2,5 g - 564,00 €
    564,00 €
    1 * 2,5 g
       564,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.