
(−)-Caryophyllene oxide 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1139-30-6
:
Formula:C₁₅H₂₄O
MDL Number:MFCD00134216
:220.356 g/mol
Melting Pt:55…62 °C
Density:0.96 g/cm³
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:3077
ADR:9,III
:GHS09
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:55 to 64 °C
- GC:≥94 %
- Specific optical rotation:-55° to -75° (20 °C, 589 nm) (c=10, EtOH)
Specification Test Results
Frequently Bought Together