Order Entry
France
ContactUsLinkComponent
Formic acid-[D2] 95% (99% D) in D₂O for NMR spectroscopy
Formic acid-[D2] 95% (99% D) in D₂O for NMR spectroscopy
Certificates
undefined
Formic acid-[D2] 95% (99% D) in D₂O for NMR spectroscopy

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • D-Enrichment:
    ≥99.0 %
  • Titration with NaOH:
    ≥94.0 %

Specification Test Results

Related Information

Frequently Bought Together