Ethyl-(S)-(+)-2-pyrrolidinone-5-carboxylate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7149-65-7
:
Formula:C₇H₁₁NO₃
MDL Number:MFCD00064497
:157.169 g/mol
Boiling Pt:176 °C (12 mmHg)
Melting Pt:47…51 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to cream Low melting solid
- Infrared spectrum:Conforms
- GC:≥98.5 %
- Specific optical rotation:-2° to -5° (20°C, 589 nm) (c=5, H2O)
Specification Test Results
Frequently Bought Together