Coating solution for DNA extraction, Pierce™
100 mL
Catalog # PIER17250
Supplier: Thermo Fisher Scientific
Coating solution for DNA extraction, Pierce™
Catalog # PIER17250
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Thermo Scientific DNA Coating Solution is an efficient method for coating DNA on the surface of microplates and microsample tubes.
- DNA Coating Solution promotes efficient immobilization of DNA onto microplates
- Hybridization efficiency is related to efficiency of DNA surface immobilization
- Facilitates high-efficiency coating of DNA onto polystryene or polypropylene surfaces
- Target DNA coated onto microplates permits speed and flexibility in probing
- DNA coated onto plastic surfaces is stabilized sufficiently to allow storage for months prior to use
- DNA Coating Solution autoclaved to inactivate DNase
: For Research Use Only. Not for use in diagnostic procedures.
- Storage Temperature:Ambient
Specifications
Specifications
- Pk:100 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



