Order Entry
France
ContactUsLinkComponent
Lugol's iodine solution USP Test Solution (TS)
Lugol's iodine solution USP Test Solution (TS)
Catalog # 85350.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Lugol's iodine solution USP Test Solution (TS)
Catalog # 85350.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
Boiling Pt:100 °C
Storage Temperature:Ambient
EINECS:0