Hygiene sponge samplers
Catalog # TSCLTS/15-B
Supplier: TSC LIMITED
Specifications
- Colour:Blue
- Description:Collection sponges in poly envelopes
- Seq:04
- Pk:200
Specifications
About this item
Sampling sponge dosed with neutralising buffer for the recovery of organisms including Listeria spp., Staphylococcus, Enterobacteriaceae such as salmonella.
Designed for the hygiene monitoring where chemical residue neutralisation is desired.
Packaging: Individual easy open stomacher pouch.