Monomeric avidin agarose, Pierce™
Supplier: Thermo Fisher Scientific
Certificates
About this item
Pierce™ Monomeric avidin agarose is immobilised onto 4% beaded agarose for purifying biotinylated proteins, peptides and other molecules.
- Non-denaturing purifies biotinylated products using mild elution conditions (2 mM free biotin)
- Reusable at least 10 times, with marginal loss in biotin binding capacity (approximately 2,5% decrease per regeneration)
- Retains biotin-binding specificity and low non specific binding of native avidin
- Binding capacity is >1,2 mg biotinylated BSA/ml resin
Frequently Bought Together