Order Entry
France
ContactUsLinkComponent
Calf alkaline phosphatase (from intestine), MP Biomedicals
Calf alkaline phosphatase (from intestine), MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Calf alkaline phosphatase (from intestine), MP Biomedicals
Supplier:  MP Biomedicals
Certificates

Specifications

  • CAS No.:
    9001-78-9
  • Source:
    Intestine
  • Species:
    Calf
  • Storage conditions:
    −20 °C
  • Form:
    Off-white powder
  • Enzyme name:
    Alkaline Phosphatase
  • Enzyme activity:
    ≥1,0 u/mg
  • pH:
    9,8

Specifications

Related Information

Frequently Bought Together