Order Entry
France
ContactUsLinkComponent
Formaldehyde dehydrogenase, MP Biomedicals
Formaldehyde dehydrogenase, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Formaldehyde dehydrogenase, MP Biomedicals
Supplier:  MP Biomedicals
Certificates

Specifications

  • CAS No.:
    9028-84-6
  • Form:
    Crystalline Powder
  • Enzyme name:
    Formaldehyde dehydrogenase

Specifications

Related Information

Frequently Bought Together