Neuraminidase (from Bacteria), MP Biomedicals
Catalog # ICNA0215173805
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:5 AU
- CAS No.:9001-67-6
- Source:Bacteria
- Seq:01
- Storage conditions:2...8 °C
- Form:White lyophilized powder
- Enzyme name:Neuraminidase
- Enzyme activity:≥50 u/mg solid
- Enzyme synonyms:Acylneuramyl hydrolase
- Molecular weight:85 kDa
- pH:6,0 - 6,5
Specifications
About this item
Neuraminidase of non-pathogenic streptococci, is prepared from the culture filtrate by Ammonium Sulfate precipitation and Ion-exchange chromatography.
- Enzyme Commission Number: E.C.3.2.1.18
- Activators: Mg2+, Co2+, Ca2+, and K+ (10mM), these activators accelerate activity 10 to 20%
- Inhibitors: Ag+, Hg+, Cu2+, and Potassium Permanganate (10mM)
- Isoelectric point (pI): 7,7 ±0,1
Unit definition: One unit causes the hydrolysis of one µmole of sialyllactose per minute at pH 6,5, 37 °C.