L-Glutamic dehydrogenase, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
For Continuous Spectrophotometric Rate determination method: 0,3 to 0,6 unit/ml of L-Glutamic dehydrogenase in cold Triethanolamine buffer.
One unit will cause the transformation of 1,0 µmole of alpha-ketoglutarate per minute at pH 7,3 and 25 °C.
LDH ≤0,01%; MDH ≤0,01%; Inhibitor: Dimethyl dicarbonate.
Specifications
- CAS No.:9029-12-3
- Form:Suspension
- Enzyme name:L-Glutamic Dehydrogenase
Specifications
Frequently Bought Together