Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Diaphorase (from
Clostridium kluyver
i), MP Biomedicals
Diaphorase (from Clostridium kluyveri), MP Biomedicals
Diaphorase (from Clostridium kluyveri), MP Biomedicals
Catalog # ICNA0215084301
Supplier:
MP Biomedicals
CAS Number:
Expand image
Diaphorase (from Clostridium kluyveri), MP Biomedicals
Catalog # ICNA0215084301
Supplier:
MP Biomedicals
CAS Number:
Specifications
Pk:
1,8 KU
CAS No.:
9001-18-7
Source:
Clostridium kluyveri
Storage conditions:
−20 °C
Enzyme name:
Diaphorase
Specifications
About this item
Product Details
Product Details